Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257386.1 | NP_001244315.1 | 6 | 934 | ||
NM_004827.2 | NP_004818.2 | 6 | 1079 | |||
XM_005263354.3 | XP_005263411.1 | 6 | 922 | |||
XM_005263355.3 | XP_005263412.1 | 6 | 1029 | |||
XM_005263356.3 | XP_005263413.1 | 6 | 805 | |||
XM_011532420.2 | XP_011530722.1 | 8 | 1193 | |||
XM_017008852.1 | XP_016864341.1 | 6 | 922 | |||
XM_017008853.1 | XP_016864342.1 | 6 | 719 | |||
GenBank mRNA | AB051855.1 | - | 6 | 630 | ||
AB056867.1 | - | 5 | 586 | |||
AF098951.2 | - | 6 | 824 | |||
AF103796.1 | - | 6 | 790 | |||
AF463519.1 | - | 5 | 594 | |||
AK002040.1 | - | 3 | 274 | |||
AK290000.1 | - | 6 | 650 | |||
AY017168.1 | - | 6 | 779 | |||
AY288307.1 | - | 3 | 244 | |||
AY289766.1 | - | 6 | 786 | |||
AY333755.1 | - | 5 | 595 | |||
AY333756.1 | - | 5 | 595 | |||
BC021281.2 | - | 6 | 1079 | 4908836 | ||
BC092408.1 | - | 6 | 934 | 6181905 |