Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lsm11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173384.1 | NP_001166855.1 | 3, 4 | 678 | ||
GenBank mRNA | BC158836.1 | - | 3, 4 | 678 | 7385359 | |
FQ223124.1 | - | 3, 4 | 714 |