Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mis12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047972.1 | NP_001041437.1 | 2 | 215 | ||
XM_006246791.2 | XP_006246853.1 | 3 | 607 | |||
XM_006246793.2 | XP_006246855.1 | 2 | 323 | |||
XM_006246794.2 | XP_006246856.1 | 2 | 446 | |||
GenBank mRNA | AY310159.2 | - | 2 | 215 |