Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slfn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768151.1 | XP_008766373.1 | 4 | 1337 |
Gene Symbol: LOC103690169 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773862.1 | XP_008772084.1 | 2 | 701 |