Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krtap13-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109325.1 | NP_001102795.1 | 1 | 479 |
Gene Symbol: LOC100363287 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002727853.3 | XP_002727899.3 | 1 | 443 |