Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC103693631 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008769095.1 | XP_008767317.1 | 377 |
Gene Symbol: Vom2r61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099508.1 | NP_001092978.1 | 1385 | |||
XM_008769096.1 | XP_008767318.1 | 2 | 125 |
Set Membership: |
G-protein coupled receptor |