Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam195b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108311.1 | NP_001101781.1 | 3 | 234 | ||
XM_006247896.2 | XP_006247958.1 | 4 | 265 | |||
XM_008768479.1 | XP_008766701.1 | 4 | 1061 | |||
GenBank mRNA | BC098826.1 | - | 3 | 241 | ||
BC158709.1 | - | 3 | 235 | 9034656 |