Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: QKI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301085.1 | NP_001288014.1 | 4 | 1049 | ||
NM_006775.2 | NP_006766.1 | 4 | 1049 | |||
NM_206853.2 | NP_996735.1 | 4 | 1049 | |||
NM_206854.2 | NP_996736.1 | 4 | 1049 | |||
NM_206855.2 | NP_996737.1 | 4 | 1049 | |||
XM_011536258.2 | XP_011534560.1 | 4 | 556 | |||
XM_011536259.2 | XP_011534561.1 | 4 | 556 | |||
XM_011536261.1 | XP_011534563.1 | 4 | 556 | |||
XM_017011502.1 | XP_016866991.1 | 4 | 556 | |||
XM_017011503.1 | XP_016866992.1 | 4 | 556 | |||
XM_017011504.1 | XP_016866993.1 | 4 | 556 | |||
GenBank mRNA | AB067798.1 | - | 4 | 1050 | ||
AB067799.1 | - | 4 | 1050 | |||
AB067800.1 | - | 4 | 1050 | |||
AB067801.1 | - | 4 | 1050 | |||
AF142422.1 | - | 4 | 609 | |||
AF272349.1 | - | 4 | 931 | |||
AK027309.1 | - | 4 | 456 | |||
AK295235.1 | - | 1 | 60 | |||
AY780788.1 | - | 4 | 486 | |||
BC012222.1 | - | 3 | 356 | |||
BC019917.2 | - | 4 | 537 | 3921310 | ||
DQ323998.1 | - | 4 | 498 |