Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITM2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021999.4 | NP_068839.1 | 5 | 840 | ||
GenBank mRNA | AF086918.1 | - | 5 | 732 | ||
AF092128.1 | - | 5 | 829 | |||
AF136973.1 | - | 5 | 829 | |||
AF152462.1 | - | 5 | 787 | |||
AF246221.1 | - | 5 | 787 | |||
AK000503.1 | - | 5 | 772 | |||
AK027411.1 | - | 5 | 771 | |||
AK130048.1 | - | 5 | 776 | |||
BC000554.2 | - | 5 | 766 | 3163436 | ||
BC016148.1 | - | 5 | 787 | 3912066 | ||
BT006863.1 | - | 5 | 617 |