Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aplp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100802.1 | NP_001094272.1 | 5 | 622 | ||
XM_008759179.1 | XP_008757401.1 | 4 | 470 | |||
GenBank mRNA | BC161904.1 | - | 5 | 622 | 8367333 | |
FQ212221.1 | - | 5 | 582 | |||
FQ213451.1 | - | 5 | 634 |