Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MAP4K4 SIRNA Assays ›
Gene Symbol:
MAP4K4
Gene Name:
mitogen-activated protein kinase kinase kinase kinase 4
Gene Aliases:
FLH21957, HEL-S-31, HGK, MEKKK4, NIK
Chromosome Location:
Chr.2: 101697703 - 101894690 on Build GRCh38
UniGene ID:
Hs.701013
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s18095
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MAP4K4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242559.1
NP_001229488.1
2
117
NM_001242560.1
NP_001229489.1
2
490
NM_004834.4
NP_004825.3
2
490
NM_145686.3
NP_663719.2
2
490
NM_145687.3
NP_663720.1
2
117
XM_005264048.2
XP_005264105.1
2
186
XM_005264050.3
XP_005264107.1
2
186
XM_005264052.3
XP_005264109.1
2
186
XM_005264057.3
XP_005264114.1
2
186
XM_005264058.3
XP_005264115.1
2
186
XM_005264059.3
XP_005264116.1
2
186
XM_005264060.3
XP_005264117.1
2
186
XM_005264062.3
XP_005264119.1
2
186
XM_005264063.3
XP_005264120.1
2
186
XM_005264064.3
XP_005264121.1
2
186
XM_005264065.3
XP_005264122.1
2
186
XM_005264066.3
XP_005264123.1
2
186
XM_005264068.3
XP_005264125.1
2
186
XM_005264069.2
XP_005264126.1
2
186
XM_005264071.2
XP_005264128.1
2
186
XM_006712865.2
XP_006712928.1
2
186
XM_006712868.2
XP_006712931.1
2
186
XM_006712869.2
XP_006712932.1
2
186
XM_017005349.1
XP_016860838.1
2
186
XM_017005350.1
XP_016860839.1
2
186
XM_017005351.1
XP_016860840.1
2
186
XM_017005352.1
XP_016860841.1
2
186
XM_017005353.1
XP_016860842.1
2
186
XM_017005354.1
XP_016860843.1
2
186
XM_017005355.1
XP_016860844.1
2
186
XM_017005356.1
XP_016860845.1
2
186
XM_017005357.1
XP_016860846.1
2
186
XM_017005358.1
XP_016860847.1
2
186
XM_017005359.1
XP_016860848.1
2
186
XM_017005360.1
XP_016860849.1
2
186
XM_017005361.1
XP_016860850.1
2
186
XM_017005362.1
XP_016860851.1
2
186
XM_017005363.1
XP_016860852.1
2
186
XM_017005364.1
XP_016860853.1
2
186
XM_017005365.1
XP_016860854.1
2
186
XM_017005366.1
XP_016860855.1
2
186
XM_017005367.1
XP_016860856.1
2
186
XM_017005368.1
XP_016860857.1
2
186
XM_017005369.1
XP_016860858.1
2
186
XM_017005370.1
XP_016860859.1
2
186
XM_017005371.1
XP_016860860.1
2
186
XM_017005372.1
XP_016860861.1
2
186
XM_017005373.1
XP_016860862.1
2
186
XM_017005374.1
XP_016860863.1
2
186
XM_017005375.1
XP_016860864.1
2
186
GenBank mRNA
AF096300.1
-
2
191
AK296396.1
-
2
186
AY212247.1
-
2
112
AY834276.1
-
2
112
DB086475.1
-
3
333
DR002278.1
-
2
499
EU794634.1
-
2
167
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_008696, NM_001252200, NM_001252201, NM_001252202, AK017168, and others.
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
signal transduction by protein phosphorylation
microvillus assembly
intracellular signal transduction
negative regulation of apoptotic process
regulation of JNK cascade
creatine kinase activity
protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
protein binding
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0