Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LY86 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004271.3 | NP_004262.1 | 4 | 420 | ||
GenBank mRNA | AB020499.1 | - | 3 | 256 | ||
AF057178.2 | - | 4 | 409 | |||
AY931019.1 | - | 4 | 396 | |||
BC038846.1 | - | 4 | 430 | 6015123 | ||
CD695164.1 | - | 4 | 484 |