Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGPS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037277.1 | NP_001032354.1 | 4 | 848 | ||
NR_036605.1 | 3 | 919 | ||||
GenBank mRNA | AB016043.1 | - | 4 | 933 | ||
AB017971.1 | - | 3 | 764 | |||
AB019036.1 | - | 4 | 974 | |||
AF057698.1 | - | 4 | 1002 | |||
AF125394.1 | - | 4 | 911 | |||
AK025139.1 | - | 4 | 996 | |||
AK293278.1 | - | 3 | 902 | |||
BC005252.1 | - | 4 | 996 | 4051481 | ||
BC067768.1 | - | 4 | 936 | 5295932 |