Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109340.1 | NP_001102810.1 | 2 | 335 | ||
XM_008761327.1 | XP_008759549.1 | 2 | 2109 |