Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOMER3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145721.1 | NP_001139193.1 | 6 | 558 | ||
NM_001145722.1 | NP_001139194.1 | 6 | 980 | |||
NM_001145724.1 | NP_001139196.1 | 4 | 418 | |||
NM_004838.3 | NP_004829.3 | 6 | 708 | |||
NR_027297.1 | 5 | 1112 | ||||
XM_006722943.1 | XP_006723006.1 | 6 | 592 | |||
XM_006722944.1 | XP_006723007.1 | 6 | 610 | |||
GenBank mRNA | AF093265.1 | - | 6 | 549 | ||
AK124450.1 | - | 5 | 1112 | |||
AK313995.1 | - | 6 | 981 | |||
AL529052.3 | - | 6 | 667 | |||
BC012113.1 | - | 6 | 650 | 4555641 | ||
BT019464.1 | - | 5 | 459 | |||
Y17573.1 | - | 5 | 459 | |||
Y18894.1 | - | 4 | 351 |