Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOMER3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145721.1 | NP_001139193.1 | 8, 9 | 904 | ||
NM_001145722.1 | NP_001139194.1 | 8, 9 | 1326 | |||
NM_001145724.1 | NP_001139196.1 | 6, 7 | 764 | |||
NM_004838.3 | NP_004829.3 | 8, 9 | 1054 | |||
NR_027297.1 | 7, 8 | 1458 | ||||
XM_006722943.1 | XP_006723006.1 | 8, 9 | 938 | |||
XM_006722944.1 | XP_006723007.1 | 8, 9 | 956 | |||
GenBank mRNA | AF093265.1 | - | 8, 9 | 895 | ||
AK124450.1 | - | 7, 8 | 1458 | |||
AK313995.1 | - | 8, 9 | 1327 | |||
BC012113.1 | - | 8, 9 | 996 | 4555641 | ||
BM915739.1 | - | 411 | ||||
BT019464.1 | - | 7, 8 | 805 | |||
Y17573.1 | - | 7, 8 | 805 | |||
Y18894.1 | - | 6, 7 | 697 | |||
Y18896.1 | - | 5, 6 | 444 |