Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008761954.1 | XP_008760176.1 | 4 | 793 | ||
XM_008761955.1 | XP_008760177.1 | 4 | 682 |