Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOMER1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004272.4 | NP_004263.1 | 6, 7 | 1751 | ||
GenBank mRNA | AF093262.1 | - | 5, 6 | 689 | ||
AK223471.1 | - | 6, 7 | 1013 | |||
AK312481.1 | - | 6, 7 | 1016 | |||
AY189940.1 | - | 2, 3 | 187 | |||
BC015502.1 | - | 6, 7 | 966 | 3904242 | ||
BC047438.1 | - | 6, 7 | 1751 | 5294849 | ||
BT009846.1 | - | 5, 6 | 689 | |||
Y17829.1 | - | 6, 7 | 764 |