Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp777 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109348.1 | NP_001102818.1 | 4 | 1145 | ||
XM_006236443.2 | XP_006236505.1 | 5 | 1385 | |||
XM_008762913.1 | XP_008761135.1 | 5 | 1174 | |||
XM_008762914.1 | XP_008761136.1 | 5 | 1199 |