Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL27RA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004843.3 | NP_004834.1 | 3 | 498 | ||
GenBank mRNA | AF053004.1 | - | 3 | 503 | ||
AF106912.1 | - | 3 | 490 | |||
AF265242.1 | - | 3 | 476 | |||
AY358876.1 | - | 3 | 476 | |||
BC028003.1 | - | 3 | 421 | 5212139 |