Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem229b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109359.1 | NP_001102829.1 | 3 | 858 | ||
XM_006240275.2 | XP_006240337.1 | 2 | 931 | |||
XM_006240276.2 | XP_006240338.1 | 2 | 927 | |||
XM_006240277.1 | XP_006240339.1 | 1 | 422 | |||
XM_008764845.1 | XP_008763067.1 | 2 | 526 | |||
GenBank mRNA | FQ211952.1 | - | 3 | 733 |