Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3BP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018009.3 | NP_001018009.2 | 8 | 1022 | ||
NM_004844.4 | NP_004835.2 | 8 | 1141 | |||
XM_011534251.2 | XP_011532553.1 | 8 | 953 | |||
XM_017007521.1 | XP_016863010.1 | 8 | 936 | |||
XM_017007522.1 | XP_016863011.1 | 9 | 1579 | |||
XM_017007523.1 | XP_016863012.1 | 8 | 1357 | |||
XM_017007524.1 | XP_016863013.1 | 8 | 889 | |||
XM_017007525.1 | XP_016863014.1 | 8 | 803 | |||
GenBank mRNA | AB005047.1 | - | 8 | 893 | ||
AK090524.1 | - | 10 | 1245 | |||
AK313167.1 | - | 8 | 1099 | |||
AL117422.1 | - | 8 | 1097 | |||
BC006169.1 | - | 8 | 996 | |||
BC010123.2 | - | 8 | 1072 | 3841593 | ||
BC023195.1 | - | 8 | 813 | |||
BX641159.1 | - | 1 | 1072 |