Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCYT1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163264.1 | NP_001156736.1 | 4 | 592 | ||
NM_001163265.1 | NP_001156737.1 | 4 | 649 | |||
NM_004845.4 | NP_004836.2 | 4 | 649 | |||
XM_017029977.1 | XP_016885466.1 | 5 | 474 | |||
GenBank mRNA | AF052510.1 | - | 4 | 476 | ||
AF148464.1 | - | 4 | 420 | |||
AK226121.1 | - | 4 | 592 | |||
AK296333.1 | - | 4 | 543 | |||
AK315323.1 | - | 4 | 649 | |||
BC045634.1 | - | 4 | 407 | 5285137 |