Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arfgap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044273.1 | NP_001037738.1 | 9 | 796 | ||
GenBank mRNA | BC099081.1 | - | 9 | 796 | 7443703 |