Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp266 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135018.1 | NP_001128490.1 | 9 | 4959 | ||
XM_008765961.1 | XP_008764183.1 | 8 | 6303 | |||
XM_008765962.1 | XP_008764184.1 | 8 | 6311 | |||
XM_008765963.1 | XP_008764185.1 | 7 | 6035 |