Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THEMIS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039477.2 | NP_001034566.1 | 4 | 1184 | ||
NM_001105556.2 | NP_001099026.1 | 6 | 2668 | |||
NM_001286113.1 | NP_001273042.1 | 7 | 2281 | |||
NM_001286115.1 | NP_001273044.1 | 6 | 2080 | |||
NM_004848.3 | NP_004839.2 | 5 | 1595 | |||
XM_005246041.3 | XP_005246098.1 | 5 | 2269 | |||
XM_006711050.1 | XP_006711113.1 | 7 | 2227 | |||
XM_011542445.2 | XP_011540747.1 | 5 | 2539 | |||
GenBank mRNA | AB050854.1 | - | 5 | 1884 | ||
AF044896.1 | - | 4 | 2321 | |||
AF323721.1 | - | 2 | 629 | |||
AK094833.1 | - | 2 | 2817 | |||
BC031655.1 | - | 5 | 1562 | |||
CR749321.1 | - | 4 | 1184 | |||
DB548309.2 | - | 1 | 620 |