Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccbl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015037.1 | NP_001015037.1 | 1124 | |||
GenBank mRNA | AY955395.1 | - | 1124 | |||
FQ218600.1 | - | 862 |