Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ugt1a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039549.1 | NP_001034638.1 | 1 | 345 | ||
GenBank mRNA | AF461734.1 | - | 1 | 354 | ||
AY435132.1 | - | 1 | 310 |