Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033073.1 | NP_001028245.1 | 1, 2 | 167 | ||
GenBank mRNA | AY623756.1 | - | 1, 2 | 167 |