Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defa6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033076.1 | NP_001028248.1 | 1 | 92 | ||
GenBank mRNA | AY623750.1 | - | 1 | 92 |