Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acsf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034951.1 | NP_001030123.1 | 9 | 1211 | ||
GenBank mRNA | BC099826.1 | - | 9 | 1211 | 7934093 |