Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc41a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037492.1 | NP_001032569.1 | 9 | 1356 | ||
XM_006236871.2 | XP_006236933.1 | 10 | 1366 | |||
XM_006236872.2 | XP_006236934.1 | 10 | 1423 | |||
XM_006236873.1 | XP_006236935.1 | 10 | 1740 | |||
XM_006236874.2 | XP_006236936.1 | 9 | 1295 | |||
XM_006236875.2 | XP_006236937.1 | 10 | 1572 | |||
XM_006236876.2 | XP_006236938.1 | 9 | 967 | |||
XM_006236877.2 | XP_006236939.1 | 9 | 1712 | |||
XM_006236878.2 | XP_006236940.1 | 9 | 1013 | |||
XM_006236879.2 | XP_006236941.1 | 9 | 1008 | |||
XM_008763134.1 | XP_008761356.1 | 10 | 1567 | |||
GenBank mRNA | BC104698.1 | - | 9 | 1356 | 7134770 |