Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defb40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037511.1 | NP_001032600.1 | 2 | 133 | ||
GenBank mRNA | AY621367.1 | - | 2 | 133 |