Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204051.1 | NP_001190980.1 | 8 | 1139 | ||
NM_004277.4 | NP_004268.3 | 8 | 1139 | |||
XM_005249483.2 | XP_005249540.1 | 8 | 1109 | |||
XM_017011512.1 | XP_016867001.1 | 7 | 988 | |||
GenBank mRNA | AF110532.1 | - | 8 | 861 | ||
AK090871.1 | - | 6 | 1468 | |||
AK291427.1 | - | 8 | 1069 | |||
AK295233.1 | - | 8 | 1122 | |||
AK302906.1 | - | 9 | 1179 | |||
AK315940.1 | - | 8 | 1050 | |||
AY358711.1 | - | 8 | 887 |