Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defb42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037532.1 | NP_001032621.1 | 2 | 168 | ||
GenBank mRNA | AY621369.1 | - | 2 | 168 |