Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defb37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037551.1 | NP_001032640.1 | 2 | 131 | ||
GenBank mRNA | AY621364.1 | - | 2 | 131 |