Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf130 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037658.1 | NP_001032747.1 | 431 | |||
GenBank mRNA | AY190520.1 | - | 431 |