Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC366747 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC088184.1 | - | 7 | 1284 |
Gene Symbol: Igh-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC086386.1 | - | 3 | 623 | ||
BC092580.1 | - | 1328 | 7368318 | |||
BC092586.1 | - | 4 | 1303 | 7366480 | ||
BC097958.1 | - | 4 | 1284 | |||
BC105825.1 | - | 5 | 1297 | 7374168 | ||
X68312.1 | - | 2 | 540 |
Gene Symbol: LOC314509 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC092582.1 | - | 1322 |
Gene Symbol: LOC678701 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC158816.1 | - | 946 |