Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004855.4 | NP_004846.4 | 11 | 1762 | ||
XM_005254795.4 | XP_005254852.1 | 9 | 1191 | |||
XM_011522235.2 | XP_011520537.1 | 11 | 1717 | |||
XM_011522236.2 | XP_011520538.1 | 10 | 1657 | |||
XM_011522237.2 | XP_011520539.1 | 10 | 1266 | |||
XM_017022730.1 | XP_016878219.1 | 10 | 1181 | |||
XM_017022731.1 | XP_016878220.1 | 11 | 1310 | |||
XM_017022732.1 | XP_016878221.1 | 9 | 1146 | |||
GenBank mRNA | AK093138.1 | - | 2 | 1503 | ||
AK223330.1 | - | 11 | 1467 | |||
AK293666.1 | - | 8 | 1378 | |||
AK314706.1 | - | 11 | 1737 | |||
D42138.1 | - | 11 | 1490 |