Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP5O | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001697.2 | NP_001688.1 | 4 | 356 | ||
GenBank mRNA | AF088071.1 | - | 2 | 151 | ||
AK124187.1 | - | 3 | 1497 | |||
AK222608.1 | - | 4 | 288 | |||
AK222962.1 | - | 4 | 283 | |||
AK311796.1 | - | 4 | 283 | |||
AV734214.1 | - | 4 | 356 | |||
BC021233.2 | - | 4 | 269 | 4126036 | ||
BC022865.1 | - | 4 | 287 | 4103001 | ||
BT019836.1 | - | 4 | 265 | |||
CD174837.1 | - | 4 | 326 | |||
CR456822.1 | - | 4 | 265 | |||
X83218.1 | - | 4 | 301 | |||
X83219.1 | - | 1 | 67 |