Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efcab12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749810.3 | XP_003749858.1 | 2 | 1412 | ||
XM_008763265.1 | XP_008761487.1 | 2 | 308 |