Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001059437.5 | XP_001059437.3 | 2 | 425 | ||
GenBank mRNA | BC158892.1 | - | 2 | 246 |