Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014243.2 | NP_055058.2 | 19 | 2718 | ||
XM_011532421.2 | XP_011530723.1 | 19 | 2750 | |||
XM_011532422.2 | XP_011530724.1 | 19 | 3249 | |||
GenBank mRNA | AB002364.1 | - | 19 | 2670 | ||
BC016451.1 | - | 2 | 158 | |||
BC130287.1 | - | 19 | 2715 | 40146318 | ||
BC132735.1 | - | 19 | 2715 | 40146757 |