Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053330.1 | NP_445782.1 | 6 | 525 | ||
GenBank mRNA | BC058459.1 | - | 6 | 529 | 6918772 | |
BC086441.1 | - | 6 | 518 | 7303531 | ||
FQ217086.1 | - | 6 | 524 | |||
FQ217504.1 | - | 6 | 523 | |||
FQ221902.1 | - | 6 | 523 | |||
FQ222392.1 | - | 6 | 524 | |||
FQ223144.1 | - | 6 | 523 | |||
FQ224800.1 | - | 6 | 524 | |||
FQ228557.1 | - | 6 | 524 | |||
M27905.1 | - | 6 | 525 | |||
X15216.1 | - | 6 | 525 |