Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gemin7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109170.1 | NP_001102640.1 | 3 | 962 | ||
XM_006228460.2 | XP_006228522.1 | 2 | 1202 | |||
XM_006228461.2 | XP_006228523.1 | 2 | 1136 |
Gene Symbol: Gemin7l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749742.2 | XP_003749790.1 | 1 | 729 |