Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casq1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159594.1 | NP_001153066.1 | 2, 3 | 528 | ||
GenBank mRNA | CA334109.1 | - | 2, 3 | 528 | ||
CB699232.1 | - | 2, 3 | 115 | |||
FQ216689.1 | - | 2, 3 | 105 | |||
FQ224875.1 | - | 3, 4 | 355 |