Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STXBP5L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308330.1 | NP_001295259.1 | 22 | 2544 | ||
NM_014980.2 | NP_055795.1 | 23 | 2616 | |||
XM_006713825.3 | XP_006713888.1 | 23 | 2786 | |||
XM_011513328.2 | XP_011511630.1 | 23 | 2591 | |||
XM_011513332.2 | XP_011511634.1 | 9 | 1093 | |||
XM_017007530.1 | XP_016863019.1 | 23 | 2547 | |||
XM_017007531.1 | XP_016863020.1 | 22 | 2529 | |||
XM_017007532.1 | XP_016863021.1 | 22 | 2478 | |||
XM_017007533.1 | XP_016863022.1 | 20 | 2204 | |||
GenBank mRNA | AB023223.1 | - | 23 | 2583 | ||
AK296544.1 | - | 22 | 2542 | |||
BC037531.2 | - | 11 | 1178 |