Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100911241 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749196.2 | XP_003749244.1 | 4 | 237 |
Gene Symbol: LOC103694201 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772077.1 | XP_008770299.1 | 4 | 246 |
Gene Symbol: Spink12l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001061351.3 | XP_001061351.1 | 4 | 237 |