Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sepsecs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128287.2 | NP_001121759.1 | 7 | 859 | ||
GenBank mRNA | BC166584.1 | - | 7 | 859 | 7443474 |