Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GDF15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004864.2 | NP_004855.2 | 2 | 550 | ||
GenBank mRNA | AB000584.1 | - | 2 | 550 | ||
AF003934.1 | - | 2 | 552 | |||
AF019770.1 | - | 2 | 535 | |||
AK225431.1 | - | 2 | 550 | |||
AK291530.1 | - | 2 | 550 | |||
BC000529.2 | - | 2 | 529 | 2961614 | ||
BC008962.2 | - | 2 | 535 | 3009858 | ||
BT019465.1 | - | 2 | 518 | |||
U51731.1 | - | 2 | 530 | |||
U88323.1 | - | 2 | 543 |